Product Details

SNP ID
rs181860697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51889376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACCCTTGCTGCCCTGCAGGCCA[C/T]GTGCCTCTAGTGAGTCTCCTGCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD33 PubMed Links

Gene Details

Gene
ANKRD33
Gene Name
ankyrin repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130015.1 629 Silent Mutation CAC,CAT H42H NP_001123487.1
NM_001304459.1 629 Silent Mutation CAC,CAT H42H NP_001291388.1
NM_001304460.1 629 UTR 5 NP_001291389.1
NM_182608.3 629 Silent Mutation CAC,CAT H177H NP_872414.3
XM_017019258.1 629 Silent Mutation CAC,CAT H186H XP_016874747.1

View Full Product Details