Product Details
- SNP ID
-
rs182620597
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:58771706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCCGCCGAGGCGCCTCTGAGAG[A/G]GCAGTACAAGGTGACCGCCTCCCCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM63B
PubMed Links
Gene Details
- Gene
- FAM63B
- Gene Name
- family with sequence similarity 63 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001040450.2 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
NP_001035540.1 |
NM_001040453.2 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
NP_001035543.1 |
XM_011521687.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_011519989.1 |
XM_011521688.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_011519990.1 |
XM_011521689.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_011519991.1 |
XM_011521690.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_011519992.1 |
XM_017022332.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_016877821.1 |
XM_017022333.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_016877822.1 |
XM_017022334.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_016877823.1 |
XM_017022335.1 |
513 |
Missense Mutation |
GAG,GGG |
E104G |
XP_016877824.1 |
- Gene
- LOC101928725
- Gene Name
- uncharacterized LOC101928725
There are no transcripts associated with this gene.
View Full Product Details