Product Details

SNP ID
hCV178259711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:145960413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGCATAAGGAGGGGCAGGCAA[T/C]GAAGGCGCCCGACCTCCGGGTTGTG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ANKRD34A PubMed Links

Gene Details

Gene
ANKRD34A
Gene Name
ankyrin repeat domain 34A
There are no transcripts associated with this gene.

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
There are no transcripts associated with this gene.

Gene
POLR3GL
Gene Name
RNA polymerase III subunit G like
There are no transcripts associated with this gene.

View Full Product Details