Product Details

SNP ID
rs182376439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50435855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAAGCCGGACTCCGTCTCAGT[A/G]GAGACTGGTGAGGGGAACCCGGGGG
Phenotype
MIM: 160793 MIM: 606802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYBPC2 PubMed Links

Gene Details

Gene
MYBPC2
Gene Name
myosin binding protein C, fast type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004533.3 240 Silent Mutation GTA,GTG V63V NP_004524.3
Gene
SPIB
Gene Name
Spi-B transcription factor
There are no transcripts associated with this gene.

View Full Product Details