Product Details

SNP ID
rs183281002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49871813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGAACAGCCTGGGACACTCACC[A/G]TCTGTACTCTCGGGGTCTGACTCAC
Phenotype
MIM: 610221 MIM: 605610 MIM: 616659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKT1S1 PubMed Links

Gene Details

Gene
AKT1S1
Gene Name
AKT1 substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098632.2 839 Silent Mutation GAC,GAT D152D NP_001092102.1
NM_001098633.3 839 Silent Mutation GAC,GAT D152D NP_001092103.1
NM_001278159.1 839 Silent Mutation GAC,GAT D152D NP_001265088.1
NM_001278160.1 839 Silent Mutation GAC,GAT D152D NP_001265089.1
NM_032375.5 839 Silent Mutation GAC,GAT D172D NP_115751.3
Gene
PNKP
Gene Name
polynucleotide kinase 3'-phosphatase
There are no transcripts associated with this gene.

Gene
TBC1D17
Gene Name
TBC1 domain family member 17
There are no transcripts associated with this gene.

View Full Product Details