Product Details

SNP ID
rs183178325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119898586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAACAACTGCAGCAACCTCAG[C/T]TGGACAATCAGGATTCCCCAAGCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC23IP PubMed Links

Gene Details

Gene
SEC23IP
Gene Name
SEC23 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007190.3 559 Missense Mutation GCT,GTT A108V NP_009121.1
XM_005269469.2 559 Missense Mutation GCT,GTT A108V XP_005269526.1
XM_005269470.2 559 Missense Mutation GCT,GTT A61V XP_005269527.1
XM_017015610.1 559 Missense Mutation GCT,GTT A61V XP_016871099.1

View Full Product Details