Product Details

SNP ID
rs183803404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:124746806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCCATTGTCTGCTGTGAAACCC[C/T]TGTAGCCATGTAGGCAGTAACAATG
Phenotype
MIM: 602583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR37 PubMed Links

Gene Details

Gene
GPR37
Gene Name
G protein-coupled receptor 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005302.3 2610 Missense Mutation AGG,GGG R521G NP_005293.1

View Full Product Details