Product Details

SNP ID
rs183804936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176371892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACTGCTCTTGGAGCTCCTCTC[C/G]TAGGCTGGAGCTCTCCTGCTTGCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 828 Intron NP_001304877.1
NM_173664.5 828 Silent Mutation TCC,TCG S244S NP_775935.1
XM_011534529.2 828 Intron XP_011532831.1
XM_011534530.2 828 Intron XP_011532832.1
XM_011534531.2 828 Intron XP_011532833.1
XM_017009372.1 828 Intron XP_016864861.1
Gene
MIR1271
Gene Name
microRNA 1271
There are no transcripts associated with this gene.

View Full Product Details