Product Details

SNP ID
rs183962296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:84125135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACAACCACTCCTTGCCGGTGCA[A/G]CTCTCGGAGAACATCTAATATTGAG
Phenotype
MIM: 610201 MIM: 615410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP162 PubMed Links

Gene Details

Gene
CEP162
Gene Name
centrosomal protein 162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286206.1 4385 Silent Mutation CTG,TTG L1307L NP_001273135.1
NM_014895.3 4385 Silent Mutation CTG,TTG L1383L NP_055710.2
XM_005248674.3 4385 Silent Mutation CTG,TTG L1345L XP_005248731.1
XM_005248678.3 4385 Intron XP_005248735.1
XM_006715380.2 4385 Silent Mutation CTG,TTG L1307L XP_006715443.1
XM_011535592.2 4385 Silent Mutation CTG,TTG L994L XP_011533894.1
XM_011535594.2 4385 Intron XP_011533896.1
XM_017010483.1 4385 Silent Mutation CTG,TTG L1345L XP_016865972.1
XM_017010484.1 4385 Silent Mutation CTG,TTG L1307L XP_016865973.1
Gene
MRAP2
Gene Name
melanocortin 2 receptor accessory protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138409.2 4385 Intron NP_612418.2
XM_011535400.2 4385 Intron XP_011533702.1
XM_017010219.1 4385 Intron XP_016865708.1
XM_017010220.1 4385 Intron XP_016865709.1
XM_017010221.1 4385 Intron XP_016865710.1

View Full Product Details