Product Details

SNP ID
rs183671693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54500555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAACATTTAGGACCAGTACATC[C/G]TGAAAAAGCCGAGATAATTAGATTC
Phenotype
MIM: 141180
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NCKAP1L PubMed Links

Gene Details

Gene
NCKAP1L
Gene Name
NCK associated protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184976.1 186 Missense Mutation CCT,CGT P29R NP_001171905.1
NM_005337.4 186 Missense Mutation CCT,CGT P79R NP_005328.2

View Full Product Details