Product Details

SNP ID
rs183728932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186588870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTGTTTGAGGTTCAGACATAT[C/T]GAGGGGATAAAAATTGGGCAAATAC
Phenotype
MIM: 600976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAT1 PubMed Links

Gene Details

Gene
FAT1
Gene Name
FAT atypical cadherin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005245.3 13677 Missense Mutation AAT,GAT N4497D NP_005236.2
XM_005262834.2 13677 Missense Mutation AAT,GAT N4509D XP_005262891.1
XM_005262835.2 13677 Missense Mutation AAT,GAT N4509D XP_005262892.1
XM_006714139.2 13677 Missense Mutation AAT,GAT N4497D XP_006714202.1

View Full Product Details