Product Details

SNP ID
rs184095805
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:17069204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCACGCACGTGCATCTCCAGG[C/T]AGCATTCCTCCTCAACCTGTGGCCC
Phenotype
MIM: 607935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PADI2 PubMed Links

Gene Details

Gene
PADI2
Gene Name
peptidyl arginine deiminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007365.2 1094 Missense Mutation TAC,TGC Y613C NP_031391.2
XM_017000148.1 1094 Missense Mutation TAC,TGC Y298C XP_016855637.1

View Full Product Details