Product Details
- SNP ID
-
rs184346776
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:149578845 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGAAGAATGAAAATCTTGAAAAT[C/T]TGGAGGAAAAAGAATATTTTGGAAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GINM1
PubMed Links
Gene Details
- Gene
- GINM1
- Gene Name
- glycoprotein integral membrane 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138785.3 |
324 |
Silent Mutation |
CTG,TTG |
L101L |
NP_620140.1 |
View Full Product Details