Product Details

SNP ID
rs184521036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149247420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCCTGACCTTTGTCTGATTCCC[A/G]TCACTTAGCATGTTGTTAAACATGT
Phenotype
MIM: 602386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF212 PubMed Links

Gene Details

Gene
ZNF212
Gene Name
zinc finger protein 212
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012256.3 Intron NP_036388.2
XM_005250055.2 Intron XP_005250112.2
XM_011516581.2 Intron XP_011514883.2
XM_011516582.1 Intron XP_011514884.1
XM_017012646.1 Intron XP_016868135.1
XM_017012647.1 Intron XP_016868136.1
XM_017012648.1 Intron XP_016868137.1

View Full Product Details