Product Details

SNP ID
rs184527844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24038073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTGTCAGCAACAGCCCTTTGTT[C/T]TGGTGACAAAAATCTGGGGAAAGAC
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322963.1 1553 Intron NP_001309892.1
NM_001322964.1 1553 Intron NP_001309893.1
NM_004116.4 1553 Intron NP_004107.1
NM_054033.3 1553 Intron NP_473374.1
XM_011532699.2 1553 Intron XP_011531001.1
XM_017003593.1 1553 Intron XP_016859082.1
XM_017003594.1 1553 Intron XP_016859083.1
XM_017003595.1 1553 Intron XP_016859084.1
XM_017003596.1 1553 Intron XP_016859085.1
Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 1553 Silent Mutation CAA,CAG Q474Q NP_001135791.1
NM_025203.2 1553 Silent Mutation CAA,CAG Q474Q NP_079479.1
XM_017005029.1 1553 Silent Mutation CAA,CAG Q474Q XP_016860518.1

View Full Product Details