Product Details

SNP ID
rs184425136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:99648374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTGACGTGCACAACCGTGCCC[A/G]AATGGTAACAATGGGCATCGTTCGC
Phenotype
MIM: 607815
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKS1B PubMed Links

Gene Details

Gene
ANKS1B
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204065.1 625 Intron NP_001190994.1
NM_001204066.1 625 Intron NP_001190995.1
NM_001204067.1 625 Intron NP_001190996.1
NM_001204068.1 625 Intron NP_001190997.1
NM_001204069.1 625 Intron NP_001190998.1
NM_001204070.1 625 Intron NP_001190999.1
NM_001204079.1 625 Intron NP_001191008.1
NM_001204080.1 625 Intron NP_001191009.1
NM_001204081.1 625 Intron NP_001191010.1
NM_020140.3 625 Intron NP_064525.1
NM_152788.4 625 Intron NP_690001.3
NM_181670.3 625 Intron NP_858056.2
XM_005269028.4 625 Intron XP_005269085.1
XM_005269029.4 625 Intron XP_005269086.1
XM_005269032.3 625 Intron XP_005269089.1
XM_006719504.3 625 Intron XP_006719567.1
XM_006719505.3 625 Intron XP_006719568.1
XM_006719506.3 625 Intron XP_006719569.1
XM_006719507.3 625 Intron XP_006719570.1
XM_006719508.3 625 Intron XP_006719571.1
XM_006719509.3 625 Intron XP_006719572.1
XM_006719510.3 625 Intron XP_006719573.1
XM_006719512.3 625 Intron XP_006719575.1
XM_006719513.3 625 Intron XP_006719576.1
XM_006719514.3 625 Intron XP_006719577.1
XM_011538571.2 625 Intron XP_011536873.1
XM_017019651.1 625 Intron XP_016875140.1
XM_017019652.1 625 Intron XP_016875141.1
XM_017019653.1 625 Intron XP_016875142.1
XM_017019654.1 625 Intron XP_016875143.1
XM_017019655.1 625 Intron XP_016875144.1
XM_017019656.1 625 Intron XP_016875145.1
XM_017019657.1 625 Intron XP_016875146.1
XM_017019658.1 625 Intron XP_016875147.1
XM_017019659.1 625 Intron XP_016875148.1
XM_017019660.1 625 Intron XP_016875149.1
XM_017019661.1 625 Intron XP_016875150.1
XM_017019662.1 625 Intron XP_016875151.1
XM_017019663.1 625 Intron XP_016875152.1
XM_017019664.1 625 Intron XP_016875153.1
XM_017019665.1 625 Intron XP_016875154.1
Gene
FAM71C
Gene Name
family with sequence similarity 71 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153364.3 625 Missense Mutation CAA,CGA Q67R NP_699195.1

View Full Product Details