Product Details
- SNP ID
-
rs184126538
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:7773831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCTGTGTTGCCAAGGCTGGTCT[C/T]GAACTCCTGCCCTCAAGTGATCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NANOGNB
PubMed Links
Gene Details
- Gene
- NANOGNB
- Gene Name
- NANOG neighbor homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145465.1 |
617 |
Nonsense Mutation |
CGA,TGA |
R183* |
NP_001138937.1 |
View Full Product Details