Product Details

SNP ID
rs184286618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72731305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCCAGGGAAATTTAGATGGTCT[A/G]TTGATCAACTAGCTGTAATAAATCC
Phenotype
MIM: 610510 MIM: 613448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BORA PubMed Links

Gene Details

Gene
BORA
Gene Name
bora, aurora kinase A activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286746.1 452 Missense Mutation ATT,GTT I135V NP_001273675.1
NM_001286747.1 452 Intron NP_001273676.1
NM_024808.3 452 Missense Mutation ATT,GTT I120V NP_079084.3
XM_006719868.3 452 Intron XP_006719931.2
Gene
MZT1
Gene Name
mitotic spindle organizing protein 1
There are no transcripts associated with this gene.

View Full Product Details