Product Details

SNP ID
rs183994350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87303060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTAGCTGAAGATTCCAACTTCA[A/G]ACCCCAAAACACTGGCTGGAAAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf95 PubMed Links

Gene Details

Gene
C16orf95
Gene Name
chromosome 16 open reading frame 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195124.1 715 Silent Mutation GTC,GTT V239V NP_001182053.1
NM_001195125.1 715 UTR 3 NP_001182054.1
NM_001256917.1 715 UTR 3 NP_001243846.1

View Full Product Details