Product Details

SNP ID
rs184015422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14409289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCCAGGGGAAAGCAACATGCCC[A/G]TGAGGCTGCTCACCCGGTGCAGGTA
Phenotype
MIM: 601211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRE5 PubMed Links

Gene Details

Gene
ADGRE5
Gene Name
adhesion G protein-coupled receptor E5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025160.2 Intron NP_001020331.1
NM_001784.4 Intron NP_001775.2
NM_078481.3 Intron NP_510966.1
XM_011528451.2 Intron XP_011526753.1
XM_017027547.1 Intron XP_016883036.1
Gene
DDX39A
Gene Name
DEAD-box helicase 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005804.3 Intron NP_005795.2
XM_006722606.3 Intron XP_006722669.1
XM_011527620.1 Intron XP_011525922.1
XM_011527621.2 Intron XP_011525923.1

View Full Product Details