Product Details

SNP ID
rs184956116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38145306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAAGCACCATGCAAGTGGGACC[A/G]TGCTCCGCCCTGGCAAATGGAGAAG
Phenotype
MIM: 604782 MIM: 600617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASH2L PubMed Links

Gene Details

Gene
ASH2L
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
There are no transcripts associated with this gene.

Gene
STAR
Gene Name
steroidogenic acute regulatory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000349.2 924 Silent Mutation CAC,CAT H220H NP_000340.2
XM_006716392.2 924 Intron XP_006716455.1

View Full Product Details