Product Details

SNP ID
rs184696635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5595501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATTATGCAAAGGGGCTTTTACT[C/T]GTATAACTTTATATGTTTATTTAGA
Phenotype
MIM: 608487 MIM: 607564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM5 PubMed Links

Gene Details

Gene
TRIM5
Gene Name
tripartite motif containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033034.2 Intron NP_149023.2
NM_033092.2 Intron NP_149083.2
NM_033093.2 Intron NP_149084.2
XM_005253183.2 Intron XP_005253240.1
XM_005253184.2 Intron XP_005253241.1
XM_006718358.2 Intron XP_006718421.1
XM_011520426.2 Intron XP_011518728.1
XM_011520427.1 Intron XP_011518729.1
XM_017018460.1 Intron XP_016873949.1
XM_017018461.1 Intron XP_016873950.1
XM_017018462.1 Intron XP_016873951.1
XM_017018463.1 Intron XP_016873952.1
Gene
TRIM6
Gene Name
tripartite motif containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003818.2 Intron NP_001003818.1
NM_001198644.1 Intron NP_001185573.1
NM_001198645.1 Intron NP_001185574.1
NM_058166.4 Intron NP_477514.1
Gene
TRIM6-TRIM34
Gene Name
TRIM6-TRIM34 readthrough
There are no transcripts associated with this gene.

View Full Product Details