Product Details
- SNP ID
-
rs184828771
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:9690389 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGTTTACTACGATAGGAAGAAG[G/T]AATGAAGGTCTTCCCACATTCAACA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF812P
PubMed Links
Gene Details
- Gene
- ZNF812P
- Gene Name
- zinc finger protein 812, pseudogene
There are no transcripts associated with this gene.
View Full Product Details