Product Details

SNP ID
rs184583395
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:28553707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCATGGGGGAGACCGTAATGA[A/G]CGTCAAAATTATCCGAAACCGCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 189 Missense Mutation AAC,AGC N32S NP_060316.1
XM_017001542.1 189 Intron XP_016857031.1

View Full Product Details