Product Details

SNP ID
rs184540632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8522467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGTATAGGGCCCGGCACCTGG[A/G]ACCATGTGTCCGAGGCTGGGGCTTG
Phenotype
MIM: 601480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1F PubMed Links

Gene Details

Gene
MYO1F
Gene Name
myosin IF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012335.3 3196 Missense Mutation CCC,TCC P1044S NP_036467.2
XM_011528024.1 3196 Missense Mutation CCC,TCC P1065S XP_011526326.1
XM_011528025.1 3196 Missense Mutation CCC,TCC P1061S XP_011526327.1
XM_011528026.1 3196 Missense Mutation CCC,TCC P1036S XP_011526328.1
XM_011528027.1 3196 Missense Mutation CCC,TCC P1020S XP_011526329.1
XM_011528028.2 3196 Intron XP_011526330.1
XM_017026821.1 3196 Missense Mutation CCC,TCC P1040S XP_016882310.1
Gene
ZNF414
Gene Name
zinc finger protein 414
There are no transcripts associated with this gene.

View Full Product Details