Product Details
- SNP ID
-
rs185188939
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:243221800 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGTTGATGACAGCGTGTTGCTTA[C/T]CCACACTACGAGACTGAAAGGAATG
- Phenotype
-
MIM: 613023
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CEP170
PubMed Links
Gene Details
- Gene
- CEP170
- Gene Name
- centrosomal protein 170
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042404.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
NP_001035863.1 |
NM_001042405.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
NP_001035864.1 |
NM_014812.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
NP_055627.2 |
XM_006711843.3 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_006711906.1 |
XM_011544334.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542636.1 |
XM_011544335.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542637.1 |
XM_011544336.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542638.1 |
XM_011544337.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542639.1 |
XM_011544338.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542640.1 |
XM_011544339.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542641.1 |
XM_011544340.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542642.1 |
XM_011544341.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542643.1 |
XM_011544342.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542644.1 |
XM_011544343.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542645.1 |
XM_011544344.2 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_011542646.1 |
XM_017002932.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858421.1 |
XM_017002933.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858422.1 |
XM_017002934.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858423.1 |
XM_017002935.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858424.1 |
XM_017002936.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858425.1 |
XM_017002937.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858426.1 |
XM_017002938.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858427.1 |
XM_017002939.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858428.1 |
XM_017002940.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858429.1 |
XM_017002941.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858430.1 |
XM_017002942.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858431.1 |
XM_017002943.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858432.1 |
XM_017002944.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858433.1 |
XM_017002945.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858434.1 |
XM_017002946.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858435.1 |
XM_017002947.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858436.1 |
XM_017002948.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858437.1 |
XM_017002949.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858438.1 |
XM_017002950.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858439.1 |
XM_017002951.1 |
527 |
Missense Mutation |
GAT,GGT |
D40G |
XP_016858440.1 |
View Full Product Details