Product Details

SNP ID
rs185564694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208239878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATCTTACCAATGGGATTGGTGG[A/G]CGTCTCCTGTCCTTTCTGGTACATG
Phenotype
MIM: 147700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDH1 PubMed Links

Gene Details

Gene
IDH1
Gene Name
isocitrate dehydrogenase (NADP(+)) 1, cytosolic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282386.1 1339 Intron NP_001269315.1
NM_001282387.1 1339 Missense Mutation CCC,TCC P326S NP_001269316.1
NM_005896.3 1339 Intron NP_005887.2

View Full Product Details