Product Details
- SNP ID
-
rs185133224
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:129177013 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTGAGATGTTGGACGCGTCCGGG[G/T]ACGAGTCCGGGCAGAAGTCCTCGAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IER5L
PubMed Links
Gene Details
- Gene
- IER5L
- Gene Name
- immediate early response 5 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_203434.2 |
1249 |
Missense Mutation |
|
|
NP_982258.2 |
View Full Product Details