Product Details

SNP ID
rs185738115
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:203798799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGGAGGATATGCATCCTTACA[A/G]CTATTTCTCAACCCCAGCCTTTCAG
Phenotype
MIM: 613513
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZC3H11A PubMed Links

Gene Details

Gene
ZC3H11A
Gene Name
zinc finger CCCH-type containing 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319238.1 Intron NP_001306167.1
NM_001319239.1 Intron NP_001306168.1
NM_014827.5 Intron NP_055642.3
XM_005245643.2 Intron XP_005245700.1
XM_005245644.3 Intron XP_005245701.1
XM_006711673.2 Intron XP_006711736.1
XM_011510199.1 Intron XP_011508501.1
XM_011510200.1 Intron XP_011508502.1
XM_011510201.1 Intron XP_011508503.1
XM_011510202.1 Intron XP_011508504.1
XM_017002956.1 Intron XP_016858445.1
XM_017002957.1 Intron XP_016858446.1
XM_017002958.1 Intron XP_016858447.1
XM_017002959.1 Intron XP_016858448.1
XM_017002960.1 Intron XP_016858449.1
XM_017002961.1 Intron XP_016858450.1
XM_017002962.1 Intron XP_016858451.1
XM_017002963.1 Intron XP_016858452.1
XM_017002964.1 Intron XP_016858453.1
XM_017002965.1 Intron XP_016858454.1

View Full Product Details