Product Details
- SNP ID
-
rs186137679
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:121852594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTCTATCAAAGAACTAGGTGAC[C/T]ACGTGCACAACTTAGTGAAGATGGG
- Phenotype
-
MIM: 608847
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FTMT
PubMed Links
Gene Details
- Gene
- FTMT
- Gene Name
- ferritin mitochondrial
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177478.1 |
640 |
Missense Mutation |
CAC,TAC |
H211Y |
NP_803431.1 |
View Full Product Details