Product Details

SNP ID
rs186133787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2179252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAGCCCCGCGACCGCCTTGAC[A/G]GGCTTGGGCGCTGTGGGCGGCGGCG
Phenotype
MIM: 612184 MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASKIN1 PubMed Links

Gene Details

Gene
CASKIN1
Gene Name
CASK interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020764.3 3881 Silent Mutation CCC,CCT P1283P NP_065815.1
Gene
TRAF7
Gene Name
TNF receptor associated factor 7
There are no transcripts associated with this gene.

View Full Product Details