Product Details

SNP ID
rs185791194
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:203765365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGATGAGATCGCACTTCCTGCAGT[A/G]GGCATTAGCCACGTCCAGGTAGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAX1 PubMed Links

Gene Details

Gene
LAX1
Gene Name
lymphocyte transmembrane adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136190.1 234 Nonsense Mutation TAG,TGG *8W NP_001129662.1
NM_001282878.1 234 Intron NP_001269807.1
NM_017773.3 234 UTR 5 NP_060243.2
XM_006711397.3 234 UTR 5 XP_006711460.1

View Full Product Details