Product Details

SNP ID
rs185908629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59639294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGAAACAGTCCACACTGACCA[C/T]TGATTATTTTGTGTTGATTCTGTAG
Phenotype
MIM: 614660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PATL1 PubMed Links

Gene Details

Gene
PATL1
Gene Name
PAT1 homolog 1, processing body mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152716.2 3404 Silent Mutation CAA,CAG Q713Q NP_689929.2
XM_005273833.4 3404 Silent Mutation CAA,CAG Q713Q XP_005273890.1
XM_005273834.1 3404 Silent Mutation CAA,CAG Q683Q XP_005273891.1
XM_011544811.2 3404 Silent Mutation CAA,CAG Q494Q XP_011543113.1
XM_017017339.1 3404 Silent Mutation CAA,CAG Q494Q XP_016872828.1

View Full Product Details