Product Details

SNP ID
rs185669877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74483387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCCAAACTTTGCAAGGAATTC[G/T]GTCCAGAGGATTACGGCGAAGAGGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC142 PubMed Links

Gene Details

Gene
CCDC142
Gene Name
coiled-coil domain containing 142
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032779.3 170 Intron NP_116168.3
Gene
TTC31
Gene Name
tetratricopeptide repeat domain 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022492.4 170 Missense Mutation GGT,TGT G36C NP_071937.4
XM_005264492.1 170 Missense Mutation GGT,TGT G36C XP_005264549.1
XM_011533035.1 170 Missense Mutation GGT,TGT G36C XP_011531337.1
XM_011533036.1 170 Missense Mutation GGT,TGT G36C XP_011531338.1
XM_011533037.1 170 Missense Mutation GGT,TGT G36C XP_011531339.1
XM_011533040.1 170 UTR 5 XP_011531342.1
XM_017004706.1 170 Missense Mutation GGT,TGT G36C XP_016860195.1
XM_017004707.1 170 Missense Mutation GGT,TGT G36C XP_016860196.1
XM_017004708.1 170 UTR 5 XP_016860197.1
XM_017004709.1 170 Intron XP_016860198.1

View Full Product Details