Product Details
- SNP ID
-
rs185688225
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:201289478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACTTCAGATGAATGTTCTCTTCC[A/G]TCAGGATCTGTATGGTGTGCTTATA
- Phenotype
-
MIM: 601763
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALS2CR12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17468277] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALS2CR12
- Gene Name
- amyotrophic lateral sclerosis 2 chromosome region candidate 12
- Gene
- CASP8
- Gene Name
- caspase 8
There are no transcripts associated with this gene.
View Full Product Details