Product Details
- SNP ID
-
rs186299011
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:447692 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGAGCCTCAAGGACGTCAAAGAC[A/G]CCGTGGTGCAGCACAGCCAGCTCGC
- Phenotype
-
MIM: 606517
MIM: 608186
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AHRR
PubMed Links
Gene Details
- Gene
- AHRR
- Gene Name
- aryl-hydrocarbon receptor repressor
There are no transcripts associated with this gene.
- Gene
- EXOC3
- Gene Name
- exocyst complex component 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007277.4 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
NP_009208.2 |
XM_011513948.2 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_011512250.1 |
XM_011513949.2 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_011512251.1 |
XM_011513950.2 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_011512252.1 |
XM_011513951.2 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_011512253.1 |
XM_017009000.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864489.1 |
XM_017009001.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864490.1 |
XM_017009002.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864491.1 |
XM_017009003.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864492.1 |
XM_017009004.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864493.1 |
XM_017009005.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864494.1 |
XM_017009006.1 |
461 |
Missense Mutation |
ACC,GCC |
T102A |
XP_016864495.1 |
- Gene
- EXOC3-AS1
- Gene Name
- EXOC3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details