Product Details

SNP ID
rs186655403
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:202881182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATCGCCGTCAGGATTGCTGCC[A/G]TCAGACAGAGCTGGCTTCTTTCTCA
Phenotype
MIM: 614522 MIM: 603417
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL12 PubMed Links

Gene Details

Gene
KLHL12
Gene Name
kelch like family member 12
There are no transcripts associated with this gene.

Gene
LOC148709
Gene Name
actin pseudogene
There are no transcripts associated with this gene.

Gene
RABIF
Gene Name
RAB interacting factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002871.4 327 Silent Mutation GAC,GAT D56D NP_002862.2

View Full Product Details