Product Details

SNP ID
rs186580848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11974037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAACGCAATCCACCTGTATCCT[G/T]GCGGACACCCGAAGAAACATACGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFB135 PubMed Links

Gene Details

Gene
DEFB135
Gene Name
defensin beta 135
There are no transcripts associated with this gene.

Gene
DEFB136
Gene Name
defensin beta 136
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033018.2 137 Missense Mutation NP_001028190.2

View Full Product Details