Product Details

SNP ID
rs186443411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127744718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATTTGGCAAATGTTATTCGGTA[C/T]TTTCCAACACAAGCTCTAAACTTTG
Phenotype
MIM: 610796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A31 PubMed Links

Gene Details

Gene
SLC25A31
Gene Name
solute carrier family 25 member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318467.1 447 Silent Mutation TAC,TAT Y93Y NP_001305396.1
NM_031291.3 447 Silent Mutation TAC,TAT Y93Y NP_112581.1
XM_011532298.1 447 Silent Mutation TAC,TAT Y93Y XP_011530600.1

View Full Product Details