Product Details

SNP ID
rs187058116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174337006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCGCTACTCGGGCCGCGCCAC[C/T]TGCGACTGCCCCAACTGCCAGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01305 PubMed Links

Gene Details

Gene
LINC01305
Gene Name
long intergenic non-protein coding RNA 1305
There are no transcripts associated with this gene.

Gene
SP9
Gene Name
Sp9 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145250.1 921 Silent Mutation ACC,ACT T307T NP_001138722.1

View Full Product Details