Product Details

SNP ID
rs186893849
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:154200209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGAGTCTCACCAAGATGAAAG[A/G]AGCAGGAACTAGAGTATACTTTGGG
Phenotype
MIM: 191030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf189 PubMed Links

Gene Details

Gene
C1orf189
Gene Name
chromosome 1 open reading frame 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010979.2 Intron NP_001010979.1
Gene
C1orf43
Gene Name
chromosome 1 open reading frame 43
There are no transcripts associated with this gene.

Gene
MIR190B
Gene Name
microRNA 190b
There are no transcripts associated with this gene.

Gene
TPM3
Gene Name
tropomyosin 3
There are no transcripts associated with this gene.

View Full Product Details