Product Details

SNP ID
rs186960267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179028127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTTCCCCAAGGCGCACATCTC[A/G]GTGAGTGACAGAGTAATTGGGAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF879 PubMed Links

Gene Details

Gene
ZNF879
Gene Name
zinc finger protein 879
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136116.1 566 Missense Mutation AGA,GGA R86G NP_001129588.1
XM_005265908.3 566 Missense Mutation AGA,GGA R86G XP_005265965.1
XM_011534550.2 566 Missense Mutation AGA,GGA R86G XP_011532852.1
XM_011534551.2 566 Missense Mutation AGA,GGA R86G XP_011532853.1

View Full Product Details