Product Details

SNP ID
rs187007388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119578339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTATAAAATCAGGTGATTCACAG[C/T]TCCAAAAATACTTATACCACAGCCC
Phenotype
MIM: 603413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIAL1 PubMed Links

Gene Details

Gene
TIAL1
Gene Name
TIA1 cytotoxic granule-associated RNA binding protein-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033925.1 Intron NP_001029097.1
NM_001323964.1 Intron NP_001310893.1
NM_001323965.1 Intron NP_001310894.1
NM_001323967.1 Intron NP_001310896.1
NM_001323968.1 Intron NP_001310897.1
NM_001323969.1 Intron NP_001310898.1
NM_001323970.1 Intron NP_001310899.1
NM_003252.3 Intron NP_003243.1
XM_017016605.1 Intron XP_016872094.1
XM_017016606.1 Intron XP_016872095.1

View Full Product Details