Product Details

SNP ID
rs186729002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28372157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGGGCGGTGGCACGGGCCCAT[G/T]GTGGGCTGCGGGTGGCCGCGGGCCC
Phenotype
MIM: 607732 MIM: 610975 MIM: 616815 MIM: 193190
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SARM1 PubMed Links

Gene Details

Gene
SARM1
Gene Name
sterile alpha and TIR motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015077.3 496 Missense Mutation TGG,TTG W42L NP_055892.2
Gene
SEBOX
Gene Name
SEBOX homeobox
There are no transcripts associated with this gene.

Gene
TMEM199
Gene Name
transmembrane protein 199
There are no transcripts associated with this gene.

Gene
VTN
Gene Name
vitronectin
There are no transcripts associated with this gene.

View Full Product Details