Product Details

SNP ID
rs187697702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24399432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCGAGGGGCCCCGAGAGAACAT[G/T]GGCAAAGCCAAGGTAAACAGCTTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NYNRIN PubMed Links

Gene Details

Gene
NYNRIN
Gene Name
NYN domain and retroviral integrase containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025081.2 504 Missense Mutation ATG,ATT M62I NP_079357.2
XM_011537016.2 504 Intron XP_011535318.1

View Full Product Details