Product Details

SNP ID
rs187524967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43217270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCTCTCTCAGCAAAATCTGTA[C/G]CTGGCTGGAAATCAAAGTCCACCAT
Phenotype
MIM: 166945
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00854 PubMed Links

Gene Details

Gene
LINC00854
Gene Name
long intergenic non-protein coding RNA 854
There are no transcripts associated with this gene.

Gene
NBR1
Gene Name
NBR1, autophagy cargo receptor
There are no transcripts associated with this gene.

Gene
TMEM106A
Gene Name
transmembrane protein 106A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291586.1 911 Missense Mutation ACC,AGC T209S NP_001278515.1
NM_001291587.1 911 Missense Mutation ACC,AGC T161S NP_001278516.1
NM_001291588.1 911 Missense Mutation ACC,AGC T162S NP_001278517.1
NM_145041.3 911 Missense Mutation ACC,AGC T209S NP_659478.1
XM_006721658.2 911 Missense Mutation ACC,AGC T209S XP_006721721.1
XM_006721659.3 911 Missense Mutation ACC,AGC T161S XP_006721722.1
XM_011524260.1 911 Missense Mutation ACC,AGC T162S XP_011522562.1
XM_017024113.1 911 Missense Mutation ACC,AGC T209S XP_016879602.1
XM_017024114.1 911 Missense Mutation ACC,AGC T161S XP_016879603.1

View Full Product Details