Product Details
- SNP ID
-
rs187986160
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:187984 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATGTGGCTTCTTAAATGACACAT[A/G]TTTTTTCTCTTCTTCATCATTAGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB128
PubMed Links
Gene Details
- Gene
- DEFB128
- Gene Name
- defensin beta 128
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037732.2 |
242 |
Missense Mutation |
CAT,TAT |
H62Y |
NP_001032821.1 |
View Full Product Details