Product Details

SNP ID
rs188025222
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:156841778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAATAGTCTGTCTCATACACGTC[A/C]CGAGTCATCCCGAAGTCTGGAAAGT
Phenotype
MIM: 147671 MIM: 191315
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INSRR PubMed Links

Gene Details

Gene
INSRR
Gene Name
insulin receptor related receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014215.2 3713 Silent Mutation CGG,CGT R1138R NP_055030.1
Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007792.1 3713 Intron NP_001007793.1
NM_001012331.1 3713 Intron NP_001012331.1
NM_002529.3 3713 Intron NP_002520.2

View Full Product Details