Product Details
- SNP ID
-
rs188024258
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18083666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTAGACTCTGCCGAGTTCTCTG[G/T]TTTTTTCTGACACTGTTCCATATTT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SAAL1
PubMed Links
Gene Details
- Gene
- SAAL1
- Gene Name
- serum amyloid A like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138421.2 |
1158 |
Missense Mutation |
ACA,CCA |
T370P |
NP_612430.2 |
View Full Product Details