Product Details

SNP ID
rs188764632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58521021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTGTCCCATGCAGCACTGTGC[A/G]ACCGACTGGAATAACCTGAAGGATG
Phenotype
MIM: 604917 MIM: 614463 MIM: 616424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT1 PubMed Links

Gene Details

Gene
CNOT1
Gene Name
CCR4-NOT transcription complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265612.1 7401 Silent Mutation GTC,GTT V2351V NP_001252541.1
NM_016284.4 7401 Silent Mutation GTC,GTT V2356V NP_057368.3
NM_206999.2 7401 Intron NP_996882.1
Gene
NDRG4
Gene Name
NDRG family member 4
There are no transcripts associated with this gene.

Gene
SETD6
Gene Name
SET domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160305.2 7401 Intron NP_001153777.1
NM_024860.3 7401 Intron NP_079136.2
XM_011523336.1 7401 Intron XP_011521638.1

View Full Product Details